Learn More
Biomatik Corporation Phospho-Trk B (Tyr705) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50996
Encompass_Preferred
Phospho-Trk B (Tyr705) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Trk B (Tyr705) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16620. Gene ID: 4915. Target Synonyms: BDNF/NT-3 growth factors receptor precursor; GP145-TrkB; GP145-TrkB/GP95-TrkB; NTRK2; Trk-B; TrkB tyrosine kinase; kinase TrkB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1208