Learn More
Biomatik Corporation Phospho-Daxx (Ser668) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51060
Encompass_Preferred
Phospho-Daxx (Ser668) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Daxx (Ser668) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UER7. Gene ID: 1616. Target Synonyms: BING2; Death domain-associated protein 6; EAP1; ETS1 associated protein 1; Fas DEATH domain- associated protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1261