Learn More
Biomatik Corporation Phospho-BCL2 (Ser70) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51090
Encompass_Preferred
Phospho-BCL2 (Ser70) Colorimetric Cell-Based ELISA Kit for detecting Phospho-BCL2 (Ser70) in researchers plate their cell line of choice. Species Reactivity: Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10415. Gene ID: 596. Target Synonyms: Apoptosis regulator Bcl-2; B-cell CLL/lymphoma 2; BCL2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1288