Learn More
Biomatik Corporation Phospho-Integrin β1 (Thr789) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51109
Encompass_Preferred
Phospho-Integrin β1 (Thr789) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Integrin β1 (Thr789) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05556. Gene ID: 3688. Target Synonyms: CD29; FNRB; Fibronectin receptor beta subunit; ITB1; Integrin VLA-4 beta subunit; Integrin beta-1 precursor; integrin beta-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1304