Learn More
Biomatik Corporation Phospho-p90 RSK (Ser380) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51136
Encompass_Preferred
Phospho-p90 RSK (Ser380) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p90 RSK (Ser380) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15418. Gene ID: 6195. Target Synonyms: 90 kDa ribosomal protein S6 kinase 1; kinase p90RSK1; KS6A1; KS6AA; MAPKAP-K1a; p90-RSK 1; pp90RSK1; Ribosomal protein S6 kinase alpha 1; Ribosomal S6 kinase 1; RPS6KA1; RSK-1; RSK1; S6K-alpha 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1326