Learn More
Biomatik Corporation Phospho-PDGFRα (Tyr713) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51142
Encompass_Preferred
Phospho-PDGFRα (Tyr713) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PDGFRα (Tyr713) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16234. Gene ID: 5175. Target Synonyms: CD31; CD31 antigen; EndoCAM; PEC1; PECA1; PECAM; PECAM1; Platelet endothelial cell adhesion molecule precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1332