Learn More
Biomatik Corporation Phospho-SP1 (Thr453) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51158
Encompass_Preferred
Phospho-SP1 (Thr453) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SP1 (Thr453) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08047. Gene ID: 6667. Target Synonyms: TSFP1; Transcription factor SP1; transcription factor Sp1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1345