Learn More
Biomatik Corporation Phospho-TNNI3 (Ser22+Ser23) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51166
Encompass_Preferred
Phospho-TNNI3 (Ser22+Ser23) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TNNI3 (Ser22+Ser23) in researchers plate their cell line of choice. Species Reactivity: Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P48787. Gene ID: 21954. Target Synonyms: Troponin I cardiac muscle; Cardiac troponin I; TNNI3; TNNC1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1351