Learn More
Biomatik Corporation Phospho-WASP (Tyr290) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51171
Encompass_Preferred
Phospho-WASP (Tyr290) Colorimetric Cell-Based ELISA Kit for detecting Phospho-WASP (Tyr290) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42768. Gene ID: 7454. Target Synonyms: IMD2; WAS; Wiskott-Aldrich syndrome protein; Wiskott-Aldrich syndrome protein homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1355