Learn More
Biomatik Corporation Phospho-NPM (Thr199) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51203
Encompass_Preferred
Phospho-NPM (Thr199) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NPM (Thr199) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06748. Gene ID: 4869. Target Synonyms: B23.1; NPM isoform 1; NPM1; Nucleolar phosphoprotein B23; Nucleolar protein NO38; nucleophosmin; nucleophosmin (nucleolar phosphoprotein B23; numatrin) Numatrin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1381