Learn More
Biomatik Corporation Phospho-SHIP1 (Tyr1021) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51218
Encompass_Preferred
Phospho-SHIP1 (Tyr1021) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SHIP1 (Tyr1021) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92835. Gene ID: 3635. Target Synonyms: Phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 1; Inositol polyphosphate-5-phosphatase of 145 kDaSIP-145; SH2 domain-containing inositol-5'-phosphatase 1; SH2 domain-containing inositol phosphatase 1; SHIP-1; p150Ship; hp51CN; INPP5D; SHIP; SHIP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1394