Learn More
Biomatik Corporation Phospho-14-3-3 ζ/δ (Thr232) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51221
Encompass_Preferred
Phospho-14-3-3 ζ/δ (Thr232) Colorimetric Cell-Based ELISA Kit for detecting Phospho-14-3-3 ζ/δ (Thr232) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P63104. Gene ID: 7534. Target Synonyms: 14-3-3 protein zeta/delta; 1433Z; 143Z; FAS; Factor activating exoenzyme S; KCIP-1; Mitochondrial import stimulation factor S1 subunit; Protein kinase C inhibitor protein-1; YWHAZ. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1396