Learn More
Biomatik Corporation Phospho-PKD2 (Ser876) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51246
Encompass_Preferred
Phospho-PKD2 (Ser876) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PKD2 (Ser876) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9BZL6. Gene ID: 25865. Target Synonyms: HSPC187; KPCD2; KPCO; PRKD2; Protein kinase C; D2 type; Protein kinase D2; Protein kinase D2 homolog; kinase PKD2; nPKC-D2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1418