Learn More
Biomatik Corporation Phospho-C/EBP-ε (Thr74) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51258
Encompass_Preferred
Phospho-C/EBP-ε (Thr74) Colorimetric Cell-Based ELISA Kit for detecting Phospho-C/EBP-ε (Thr74) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q15744. Gene ID: 1053. Target Synonyms: C/EBP epsilon; CCAAT/enhancer binding protein epsilon; CEBPE. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1430