Learn More
Biomatik Corporation Phospho-Calmodulin (Thr79+Ser81) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51260
Encompass_Preferred
Phospho-Calmodulin (Thr79+Ser81) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Calmodulin (Thr79+Ser81) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P62158. Gene ID: 801; 805; 808. Target Synonyms: CALM; CALM1; CALM2; CALM3; CAM; CAM1; CAM2; CAM3; CAMB; CAMC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1432