Learn More
Biomatik Corporation Phospho-C-RAF (Ser43) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51285
Encompass_Preferred
Phospho-C-RAF (Ser43) Colorimetric Cell-Based ELISA Kit for detecting Phospho-C-RAF (Ser43) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P04049. Gene ID: 5894. Target Synonyms: C-RAF; C-Raf; CRAF; kinase Raf1; Protein kinase raf 1; RAF proto-oncogene serine/threonine-protein kinase; Raf proto-oncogene serine/threonine-protein kinase; RAF-1; Raf-1; RAF1; Similar to murine leukemia viral (V-raf-1) oncogene homolog 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1453