Learn More
Biomatik Corporation Phospho-CRYAB (Ser45) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51287
Encompass_Preferred
Phospho-CRYAB (Ser45) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CRYAB (Ser45) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P02511. Gene ID: 1410. Target Synonyms: Alpha crystallin B chain; Alpha(B)-crystallin; CRYA2; P23; Rosenthal fiber component; crystallin; alphaB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1455