Learn More
Biomatik Corporation Phospho-Histone H1 (Thr17) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51306
Encompass_Preferred
Phospho-Histone H1 (Thr17) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Histone H1 (Thr17) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P16401; P16402; P10412. Gene ID: 3009; 3007; 3008. Target Synonyms: Histone H1.5; Histone H1a; HIST1H1B; H1F5; Histone H1.3; Histone H1c; HIST1H1D; H1F3; Histone H1.4; Histone H1b; HIST1H1E; H1F4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1472