Learn More
Biomatik Corporation Phospho-p57 Kip2 (Thr310) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51309
Encompass_Preferred
Phospho-p57 Kip2 (Thr310) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p57 Kip2 (Thr310) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49918. Gene ID: 1028. Target Synonyms: CDKN1C; CDN1C; CDNC; Cyclin-dependent kinase inhibitor 1C; Cyclin-dependent kinase inhibitor p57; KIP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1475