Learn More
Biomatik Corporation Phospho-Smad3 (Ser208) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51319
Encompass_Preferred
Phospho-Smad3 (Ser208) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Smad3 (Ser208) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P84022. Gene ID: 4088. Target Synonyms: JV15-2; MAD-3; MADH3; Mad3; Mothers against DPP homolog 3; Mothers against decapentaplegic homolog 3; SMAD 3; Smad 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1483