Learn More
Biomatik Corporation Phospho-RIPK2 (Ser176) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51643
Encompass_Preferred
Phospho-RIPK2 (Ser176) Colorimetric Cell-Based ELISA Kit for detecting Phospho-RIPK2 (Ser176) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O43353. Gene ID: 8767. Target Synonyms: CARD-containing IL-1 beta ICE-kinase; CARD-containing interleukin-1 beta converting enzyme associated kinase; CARDIAK; kinase RIPK2; Receptor-interacting protein 2; Receptor-interacting serine/threonine protein kinase 2; RICK; RIP-2; RIP-like interacting CLARP kinase; RIP2; RIPK2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9095