Learn More
Biomatik Corporation Phospho-GTPase Activating Protein (Ser387) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51330
Encompass_Preferred
Phospho-GTPase Activating Protein (Ser387) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GTPase Activating Protein (Ser387) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9H0H5. Gene ID: 29127. Target Synonyms: GTPase activating protein; GTPase activating protein ID-GAP; MGCRACGAP variant alpha; MGCRACGAP variant beta; RAC GTPase- activating protein 1; RAC GTPase-activating protein; RACGAP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1494