Learn More
Biomatik Corporation Phospho-IRAK1 (Thr100) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51343
Encompass_Preferred
Phospho-IRAK1 (Thr100) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IRAK1 (Thr100) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P51617. Gene ID: 3654. Target Synonyms: IL1RAK; IRAK; IRAK-1; Interleukin-1 receptor-associated kinase 1; Pelle-like protein kinase; kinase IRAK1; mPLK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1507