Learn More
Biomatik Corporation Phospho-KIF11/Eg5 (Thr926) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51347
Encompass_Preferred
Phospho-KIF11/Eg5 (Thr926) Colorimetric Cell-Based ELISA Kit for detecting Phospho-KIF11/Eg5 (Thr926) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52732. Gene ID: 3832. Target Synonyms: Eg5; KIFL1; KNSL1; Kinesin-like protein 1; Kinesin-like protein KIF11; Kinesin-like spindle protein HKSP; Kinesin-related mitotic motor protein; Kinesin-related motor protein Eg5; TRIP5; Thyroid receptor interacting protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1510