Learn More
Biomatik Corporation Phospho-Adrenergic Receptor β2 (Ser346) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51356
Encompass_Preferred
Phospho-Adrenergic Receptor β2 (Ser346) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Adrenergic Receptor β2 (Ser346) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P07550. Gene ID: 154. Target Synonyms: Beta-2 adrenergic receptor; Beta-2 adrenoceptor; Beta-2 adrenoreceptor; ADRB2; ADRB2R; B2AR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1519