Learn More
Biomatik Corporation Phospho-FOXO4 (Thr451) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51360
Encompass_Preferred
Phospho-FOXO4 (Thr451) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FOXO4 (Thr451) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P98177. Gene ID: 4303. Target Synonyms: AFX; AFX1; Afxh; Forkhead box protein O4; MLLT7; Putative forkhead domain AFX1; putative fork head domain AFX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1521