Learn More
Biomatik Corporation Phospho-NEK9 (Thr210) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51363
Encompass_Preferred
Phospho-NEK9 (Thr210) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NEK9 (Thr210) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8TD19. Gene ID: 91754. Target Synonyms: KIAA1995; NERCC; NIMA-related kinase 8; Nek8; Nercc1 kinase; NimA-related protein kinase 9; Serine/threonine-protein kinase Nek9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1523