Learn More
Biomatik Corporation Phospho-NIPA (Ser354) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51365
Encompass_Preferred
Phospho-NIPA (Ser354) Colorimetric Cell-Based ELISA Kit for detecting Phospho-NIPA (Ser354) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q86WB0. Gene ID: 51530. Target Synonyms: Nuclear interacting partner of ALK; Nuclear-interacting partner of ALK; ZC3HC1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1525