Learn More
Biomatik Corporation Phospho-TISB (Ser92) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51372
Encompass_Preferred
Phospho-TISB (Ser92) Colorimetric Cell-Based ELISA Kit for detecting Phospho-TISB (Ser92) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q07352. Gene ID: 677. Target Synonyms: BERG36; Butyrate response factor 1; EGF-response factor 1; ERF-1; ERF1; TIS11B; TIS11B protein; TISB; ZFP36L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1532