Learn More
Biomatik Corporation Phospho-SNAI1 (Ser246) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51382
Encompass_Preferred
Phospho-SNAI1 (Ser246) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SNAI1 (Ser246) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O95863. Gene ID: 6615. Target Synonyms: SNAH; SNAI; Sna; Snail; zinc finger protein SNAI1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1540