Learn More
Biomatik Corporation Phospho-SHP-2 (Tyr580) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50989
Encompass_Preferred
Phospho-SHP-2 (Tyr580) Colorimetric Cell-Based ELISA Kit for detecting Phospho-SHP-2 (Tyr580) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q06124. Gene ID: 5781. Target Synonyms: PTN11; PTP-1D; PTP-2C; PTP2C; PTPN11; Protein-tyrosine phosphatase 2C; Protein-tyrosine phosphatase; non-receptor type 11; SH-PTP2; SH-PTP3; SHPTP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8997