Learn More
Biomatik Corporation Phospho-CPI17α (Thr38) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51424
Encompass_Preferred
Phospho-CPI17α (Thr38) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CPI17α (Thr38) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q96A00. Gene ID: 94274. Target Synonyms: Protein phosphatase 1 regulatory subunit 14A; 17 kDa PKC-potentiated inhibitory protein of PP1; CPI-17; PPP1R14A; CPI17; PPP1INL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1574