Learn More
Biomatik Corporation Phospho-GAB1 (Tyr627) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51458
Encompass_Preferred
Phospho-GAB1 (Tyr627) Colorimetric Cell-Based ELISA Kit for detecting Phospho-GAB1 (Tyr627) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q13480. Gene ID: 2549. Target Synonyms: GRB2-associated binder 1; GRB2-associated-binding protein 1; growth factor receptor bound protein 2-associated protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1599