Learn More
Biomatik Corporation Phospho-ICAM-1 (Tyr512) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51478
Encompass_Preferred
Phospho-ICAM-1 (Tyr512) Colorimetric Cell-Based ELISA Kit for detecting Phospho-ICAM-1 (Tyr512) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P05362. Gene ID: 3383. Target Synonyms: Intercellular adhesion molecule 1; ICAM-1; Major group rhinovirus receptor; CD54; ICAM1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1616