Learn More
Biomatik Corporation Phospho-Lck (Tyr393) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51496
Encompass_Preferred
Phospho-Lck (Tyr393) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Lck (Tyr393) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P06239. Gene ID: 3932. Target Synonyms: kinase Lck; LCK; LSK; LSK-T; lymphocyte-specific protein tyrosine kinase; P56-LCK; T cell-specific protein-tyrosine kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1633