Learn More
Biomatik Corporation Phospho-MKK6 (Ser207) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51510
Encompass_Preferred
Phospho-MKK6 (Ser207) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MKK6 (Ser207) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52564. Gene ID: 5608. Target Synonyms: Dual specificity mitogen-activated protein kinase kinase 6; kinase MKK6; MAP kinase kinase 6; MAP2K6; MAPK/ERK kinase 6; MAPKK 6; MEK6; Mitogen-activated protein kinase kinase 6; MP2K6; PRKMK6; SAPKK3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1646