Learn More
Biomatik Corporation Phospho-p21 Cip1 (Thr145) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51527
Encompass_Preferred
Phospho-p21 Cip1 (Thr145) Colorimetric Cell-Based ELISA Kit for detecting Phospho-p21 Cip1 (Thr145) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P38936. Gene ID: 1026. Target Synonyms: CAP20; CDK-interacting protein 1; CDKN1; CDKN1A; CDN1A; CIP1; Cyclin-dependent kinase inhibitor 1; MDA-6; MDA6; Melanoma differentiation associated protein; Melanoma differentiation associated protein 6; p21; PIC1; SDI1; WAF1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1661