Learn More
Biomatik Corporation Phospho-AurA (Thr288) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51089
Encompass_Preferred
Phospho-AurA (Thr288) Colorimetric Cell-Based ELISA Kit for detecting Phospho-AurA (Thr288) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O14965. Gene ID: 6790. Target Synonyms: AIK; ARK1; Aurora-A; Aurora-family kinase 1; Aurora-related kinase 1; Aurora/IPL1-related kinase 1; AYK1; Breast-tumor-amplified kinase; BTAK; IAK1; Ipl1- and aurora-related kinase 1; kinase AurA; Serine/threonine kinase 15; Serine/threonine kinase 6; Serine/threonine kinase Ayk1; STK15; STK6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9012