Learn More
Biomatik Corporation Phospho-MNK1 (Thr255) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51632
Encompass_Preferred
Phospho-MNK1 (Thr255) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MNK1 (Thr255) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9BUB5. Gene ID: 8569. Target Synonyms: kinase Mnk1; Map kinase interacting kinase; MAP kinase signal-integrating kinase 1; MAP kinase-interacting serine/threonine kinase 1; MKNK1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1746