Learn More
Biomatik Corporation Phospho-Tau (Thr231) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51588
Encompass_Preferred
Phospho-Tau (Thr231) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Tau (Thr231) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P10636. Gene ID: 4137. Target Synonyms: MAPT; Microtubule-associated protein tau; MTBT1; Neurofibrillary tangle protein; Paired helical filament-tau; PHF-tau. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1712