Learn More
Biomatik Corporation Phospho-nNOS (Ser852) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51130
Encompass_Preferred
Phospho-nNOS (Ser852) Colorimetric Cell-Based ELISA Kit for detecting Phospho-nNOS (Ser852) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P29475. Gene ID: 4842. Target Synonyms: bNOS; BNOS; Constitutive NOS; N-NOS; NC-NOS; Neuronal NOS; Nitric-oxide synthase brain; NOS type I; NOS1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9019