Learn More
Biomatik Corporation Phospho-FLT3 (Tyr842) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51623
Encompass_Preferred
Phospho-FLT3 (Tyr842) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FLT3 (Tyr842) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P36888. Gene ID: 2322. Target Synonyms: Tyrosine-protein kinase receptor FLT3; Stem cell tyrosine kinase 1; STK-1; CD135 antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1739