Learn More
Biomatik Corporation Phospho-PAK3 (Ser154) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51638
Encompass_Preferred
Phospho-PAK3 (Ser154) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PAK3 (Ser154) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75914. Gene ID: 5063. Target Synonyms: p21-activated kinase 3; PAK-3; Beta-PAK; CDC42/RAC effector kinase PAK-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1751