Learn More
Biomatik Corporation Phospho-Cofilin (Ser3) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51420
Encompass_Preferred
Phospho-Cofilin (Ser3) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Cofilin (Ser3) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P23528. Gene ID: 3716. Target Synonyms: Cofilin-1; Cofilin; non-muscle isoform; 18 kDa phosphoprotein; p18; CFL1; CFL. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9056