Learn More
Biomatik Corporation Phospho-FOS (Ser32) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51649
Encompass_Preferred
Phospho-FOS (Ser32) Colorimetric Cell-Based ELISA Kit for detecting Phospho-FOS (Ser32) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P01100. Gene ID: 2353. Target Synonyms: Cellular oncogene fos; FOS; G0/G1 switch regulatory protein 7; G0S7; Proto-oncogene protein c-fos. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1758