Learn More
Biomatik Corporation Phospho-PYK2 (Tyr579) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51661
Encompass_Preferred
Phospho-PYK2 (Tyr579) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PYK2 (Tyr579) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q14289. Gene ID: 2185. Target Synonyms: CADTK; CAK beta; Calcium-dependent tyrosine kinase; Cell adhesion kinase beta; FADK 2; FAK2; Focal adhesion kinase 2; kinase Pyk2; Proline-rich tyrosine kinase 2; Protein tyrosine kinase 2 beta; PTK2B; RAFTK; Related adhesion focal tyrosine kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1769