Learn More
Biomatik Corporation Phospho-Gab2 (Tyr643) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51675
Encompass_Preferred
Phospho-Gab2 (Tyr643) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Gab2 (Tyr643) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9UQC2. Gene ID: 9846. Target Synonyms: GRB2-associated-binding protein 2; Growth factor receptor bound protein 2-associated protein 2; GRB2-associated binder 2; pp100; KIAA0571. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1783