Learn More
Biomatik Corporation Phospho-MAPKAPK2 (Ser272) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51677
Encompass_Preferred
Phospho-MAPKAPK2 (Ser272) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MAPKAPK2 (Ser272) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49137. Gene ID: 9261. Target Synonyms: kinase MAPKAPK2; MAP kinase-activated protein kinase 2; MAPK-activated protein kinase 2; MAPK2; MAPKAP kinase 2; MAPKAPK-2; MAPKAPK2; RPS6KC1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1785