Learn More
Biomatik Corporation Phospho-PSEN1 (Ser357) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51683
Encompass_Preferred
Phospho-PSEN1 (Ser357) Colorimetric Cell-Based ELISA Kit for detecting Phospho-PSEN1 (Ser357) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P49768. Gene ID: 5663. Target Synonyms: AD3; AD3H; presenilin 1; PS-1; PS1; PSN1; PSNL1; S182 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1791