Learn More
Biomatik Corporation Phospho-RFWD2 (Ser387) Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA51685
Encompass_Preferred
Phospho-RFWD2 (Ser387) Colorimetric Cell-Based ELISA Kit for detecting Phospho-RFWD2 (Ser387) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q8NHY2. Gene ID: 64326. Target Synonyms: Constitutive photomorphogenesis protein 1 homolog; COP1; hCOP1; RING finger and WD repeat domain protein 2; Ubiquitin- protein ligase COP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-1793